Invitrogen
Manufacturer Code:MA515080
Catalog # PIMA515080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.414.5 |
Immunogen | Synthetic peptide corresponding to residues at the carboxy terminus of human IKKß protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B kinase 2; I-kappa-B kinase 2 I-kappa-B-kinase beta IKK-B inhibitor of nuclear factor kappa-B kinase subunit beta nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B-kinase beta; IKK-B; IKK-beta; IKK2; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta; Serine/threonine protein kinase IKBKB
Gene Aliases: AI132552; AIM-1; IKBKB; IKK-2; IKK-beta; IKK2; IKK[b]; IKKB; IKKbeta; IMD15; NFKBIKB
UniProt ID: (Human) O14920, (Mouse) O88351, (Rat) Q9QY78
Entrez Gene ID: (Human) 3551, (Mouse) 16150, (Rat) 84351
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.