Novus Biologicals
Manufacturer Code:NBP233214
Catalog # NBP233214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10A9B6 |
Immunogen | Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.10 I-kappa-B kinase 2 I-kappa-B-kinase beta IkBKB IKK2FLJ38368 IKK-B IKK-betaFLJ36218 IKKBFLJ40509 inhibitor of kappa light polypeptide gene enhancer in B-cells kinase beta inhibitor of nuclear factor kappa-B kinase subunit beta MGC131801 NFKBIKBFLJ33771 Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2; I-kappa-B-kinase beta; IKK-B; IKK2; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta; Serine/threonine protein kinase IKBKB
Gene Aliases: IKBKB; IKK-beta; IKK2; IKKB; IMD15; NFKBIKB
UniProt ID: (Human) O14920
Entrez Gene ID: (Human) 3551
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.