Invitrogen
Manufacturer Code:PA579456
Catalog # PIPA579456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human IKK beta recombit protein (Position: E398-S756). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B kinase 2; I-kappa-B-kinase beta; IKK-B; IKK2; Inhibitor of nuclear factor kappa-B kinase subunit beta; Inhibitor of nuclear factor kappa-B kinase subunit beta I-kappa-B-kinase beta IKK-B IKK-beta IkBKB I-kappa-B kinase 2 IKK2 Nuclear factor NF-kappa-B inhibitor kinase beta NFKBIKB IKBKB IKKB; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta; Serine/threonine protein kinase IKBKB
Gene Aliases: IKBKB; IKK-beta; IKK2; IKKB; IMD15; NFKBIKB
UniProt ID: (Human) O14920
Entrez Gene ID: (Human) 3551
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.