Invitrogen
Manufacturer Code:PA517804
Catalog # PIPA517804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues at the carboxy terminus of human IKKß |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: I-kappa-B kinase 2; I-kappa-B kinase 2 I-kappa-B-kinase beta IKK-B inhibitor of nuclear factor kappa-B kinase subunit beta nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B-kinase beta; IKK-B; IKK2; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta; Serine/threonine protein kinase IKBKB
Gene Aliases: IKBKB; IKK-beta; IKK2; IKKB; IMD15; NFKBIKB
UniProt ID: (Human) O14920
Entrez Gene ID: (Human) 3551
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.