Invitrogen
Manufacturer Code:PA517803
Catalog # PIPA517803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the 20 amino-terminal residues of human IKKa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; I kappa B kinase 1; i kappa-B kinase alpha; I-kappa-B kinase 1; I-kappa-B kinase 1 I-kappa-B kinase alpha I-kappa-B kinase-alpha IKK-a kinase IkB kinase alpha subunit Nuclear factor NFkappaB inhibitor kinase alpha TCF-16 conserved helix-loop ubiquitous kinase ikappaB kinase inhibitor of nuclear factor kappa-B kinase subunit alpha nuclear factor NF-kappa-B inhibitor kinase alpha transcription factor 16; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkappaB kinase alpha; IkB kinase alpha subunit; ikBKA; IKK alpha; IKK-A; IKK-a kinase; IKK-alpha; IKK1; Inhibitor of nuclear factor kappa-B kinase subunit alpha; NFKBIKA; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NFkappaB inhibitor kinase alpha; TCF-16; Transcription factor 16
Gene Aliases: AI256658; CHUK; Chuk1; Fbx24; Fbxo24; IKBKA; IKK-alpha; IKK1; IKKA; NFKBIKA; TCF16
UniProt ID: (Bovine) Q95KV1, (Human) O15111
Entrez Gene ID: (Bovine) 281073, (Human) 1147, (Mouse) 12675, (Rat) 309361
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.