Novus Biologicals
Manufacturer Code:NBP299049C
Catalog # NB083950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human IKK alpha. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; conserved helix-loop-helix ubiquitous kinaseIKBKA EC 2.7.11 EC 2.7.11.10 IkappaB kinase I-kappa-B kinase 1 I-kappa-B kinase alpha IkBKA IKK1I-kappa-B kinase-alpha IKK-A IKKAIkB kinase alpha subunit IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha NFKBIKAIKK-a kinase Nuclear factor NFkappaB inhibitor kinase alpha Nuclear factor NF-kappa-B inhibitor kinase alpha TCF16 TCF-16 Transcription factor 16; I-kappa-B kinase 1; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IKK-a kinase; IKK1; Inhibitor of nuclear factor kappa-B kinase subunit alpha; NFKBIKA; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NFkappaB inhibitor kinase alpha; TCF-16; Transcription factor 16
Gene Aliases: CHUK; IKBKA; IKK-alpha; IKK1; IKKA; NFKBIKA; TCF16
UniProt ID: (Human) O15111
Entrez Gene ID: (Human) 1147
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.