Novus Biologicals
Manufacturer Code:NBP30330820UL
Catalog # NB094719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human IKEPP (NP_079067.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ22756 IKEPPPDZK2 intestinal and kidney enriched PDZ protein Intestinal and kidney-enriched PDZ protein Na(+)/H(+) exchange regulatory cofactor NHE-RF4 Na/Pi cotransporter C-terminal-associated protein 2 naPi-Cap2 Natrium-phosphate cotransporter IIa C-terminal-associated protein 2 NHERF4 NHERF-4 PDZ domain containing 2 PDZ domain containing 3 PDZ domain-containing protein 2 PDZ domain-containing protein 3 Sodium-hydrogen exchanger regulatory factor 4; intestinal and kidney enriched PDZ protein; Intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; Na/Pi cotransporter C-terminal-associated protein 2; naPi-Cap2; Natrium-phosphate cotransporter IIa C-terminal-associated protein 2; NHERF-4; PDZ domain containing 2; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Sodium-hydrogen exchanger regulatory factor 4
Gene Aliases: DLNB27; IKEPP; NHERF4; PDZD3; PDZK2
UniProt ID: (Human) Q86UT5
Entrez Gene ID: (Human) 79849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.