Novus Biologicals
Manufacturer Code:NBP176788
Catalog # NBP176788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human IKAP. The immunogen is located within the last 50 amino acids of IKAP. Amino Acid Squence: PPKINRRTQWKLSLLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DYS elongator complex protein 1 ELP1DKFZp781H1425 FD FLJ12497 IKAPdysautonomia (Riley-Day syndrome hereditary sensory autonomic neuropathy typeIII) IkappaB kinase complex-associated protein IKI3 IKK complex-associated protein inhibitor of kappa light polypeptide gene enhancer in B-cells kinasecomplex-associated protein p150 TOT1; elongator acetyltransferase complex subunit 1; Elongator complex protein 1; ELP1; IkappaB kinase complex-associated protein; IKK complex-associated protein; p150
Gene Aliases: DYS; ELP1; FD; IKAP; IKBKAP; IKI3; TOT1
UniProt ID: (Human) O95163
Entrez Gene ID: (Human) 8518
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.