Novus Biologicals
Manufacturer Code:NBP132322
Catalog # NBP132322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human IP6K2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:1D-myo-inositol-hexakisphosphate phosphotransferase; EC 2.7.4.21 IHPK2ATP:1D-myo-inositol-hexakisphosphate phosphotransferase inositol hexakisphosphate kinase 2 inositol hexaphosphate kinase 2 InsP6 kinase 2 P(i)-uptake stimulator pi uptake stimulator PIUS; Inositol hexakisphosphate kinase 2; inositol hexaphosphate kinase 2; InsP6 kinase 2; P(i)-uptake stimulator; pi uptake stimulator; PiUS
Gene Aliases: IHPK2; IP6K2; PIUS; TCCCIA00113
UniProt ID: (Human) Q9UHH9
Entrez Gene ID: (Human) 51447
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.