Invitrogen
Manufacturer Code:MA537541
Catalog # PIMA537541
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 363CT4.1.6 |
Immunogen | KLH conjugated synthetic peptide selected from the C-terminal region of human IHH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BDA1 HHG2 HHG-2 IHH indian hedgehog Indian hedgehog homolog indian hedgehog protein Indian hedgehog protein C-product Indian hedgehog protein N-product; HHG-2; IHH; Indian hedgehog homolog; Indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product
Gene Aliases: BDA1; HHG2; IHH
UniProt ID: (Human) Q14623
Entrez Gene ID: (Human) 3549
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.