Invitrogen
Manufacturer Code:PA593107
Catalog # PIPA593107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 202-411 of human IHH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HHG-2; IHH; Indian hedgehog homolog; Indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product
Gene Aliases: BDA1; HHG2; IHH
UniProt ID: (Human) Q14623, (Mouse) P97812
Entrez Gene ID: (Human) 3549, (Mouse) 16147, (Rat) 84399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.