Novus Biologicals
Manufacturer Code:NBP214841SS
Catalog # NBP214841SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 41N3D3 |
Immunogen | A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BL2 cell adhesion molecule 1 IgSF4 IGSF4ADKFZp686F1789 IGSF4immunoglobulin superfamily member 4D variant 2 Immunoglobulin superfamily member 4 immunoglobulin superfamily member 4 Necl-2 NECL2MGC149785 nectin-like 2 Nectin-like protein 2 RA175 sgIGSF Spermatogenic immunoglobulin superfamily ST17 sTSLC-1 Synaptic cell adhesion molecule SynCAM synCAM1 SYNCAMMGC51880 TSLC1 TSLC-1 TSLC1/Nectin-like 2/IGSF4 Tumor suppressor in lung cancer 1SYNCAM1; CADM1; Cell adhesion molecule 1; IgSF4; Immunoglobulin superfamily member 4; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; NECL-2; nectin-like 2; Nectin-like protein 2; SgIgSF; Spermatogenic immunoglobulin superfamily; Synaptic cell adhesion molecule; SynCAM; truncated CADM1 protein; TSLC-1; TSLC1/Nectin-like 2/IGSF4; Tumor suppressor in lung cancer 1
Gene Aliases: BL2; CADM1; IGSF4; IGSF4A; Necl-2; NECL2; RA175; sgIGSF; ST17; sTSLC-1; SYNCAM; synCAM1; TSLC1
UniProt ID: (Human) Q9BY67
Entrez Gene ID: (Human) 23705
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.