Proteintech
Manufacturer Code:136881AP150UL
Catalog # 50-172-8937
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IGJ Fusion Protein Ag4559 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IGCJ IGJ Immunoglobulin J chain JCH; IgJ chain; Immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; immunoglobulin joining gene (not J region); J chain; Joining chain of multimeric IgA and IgM
Gene Aliases: 9530090F24Rik; AI323815; IGCJ; IGJ; JCH; JCHAIN
UniProt ID: (Human) P01591, (Mouse) P01592
Entrez Gene ID: (Human) 3512, (Mouse) 16069
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.