Invitrogen
Manufacturer Code:PA579453
Catalog # PIPA579453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human IGFBP5 recombit protein (Position: A60-E272). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP-5; IGF-binding protein 5; IGFBP-5; insulin-like growth factor binding protein 5; Insulin-like growth factor-binding protein 5; Insulin-like growth factor-binding protein 5 IBP-5 IGF-binding protein 5 IGFBP-5 IGFBP5 IBP5
Gene Aliases: IBP5; IGFBP5
UniProt ID: (Human) P24593
Entrez Gene ID: (Human) 3488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.