Invitrogen
Manufacturer Code:PA570003
Catalog # PIPA570003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human IGFBP5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP 5 IBP-5 IBP5 IBP5 IGF binding protein 5 IGF BP5 IGF-binding protein 5 IGFBP 5 IGFBP-5 IGFBP5 Insulin Like Growth Factor Binding Protein 5 Insulin-like growth factor-binding protein 5; IBP-5; IGF-binding protein 5; IGFBP-5; insulin-like growth factor binding protein 5; Insulin-like growth factor-binding protein 5
Gene Aliases: AI256729; AW208790; IBP5; IGF-BP5; IGFBP-5; IGFBP-5P; IGFBP5
UniProt ID: (Pig) Q28985, (Human) P24593, (Bovine) Q05717, (Mouse) Q07079, (Rat) P24594
Entrez Gene ID: (Pig) 397182, (Goat) 100860950, (Sheep) 443133, (Dog) 610316, (Horse) 100034063, (Rabbit) 100348897, (Human) 3488, (Bovine) 404185, (Mouse) 16011, (Rat) 25285, (Guinea pig) 100724427
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.