Invitrogen
Manufacturer Code:PA547775
Catalog # PIPA547775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse IGFBP-5 Val15-Glu271 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP 5 IBP-5 IBP5 IBP5 IGF binding protein 5 IGF BP5 IGF-binding protein 5 IGFBP 5 IGFBP-5 IGFBP5 Insulin Like Growth Factor Binding Protein 5 Insulin-like growth factor-binding protein 5; IBP-5; IGF-binding protein 5; Insulin-like growth factor-binding protein 5
Gene Aliases: AI256729; AW208790; IGFBP-5; IGFBP-5P; Igfbp5
UniProt ID: (Mouse) Q07079
Entrez Gene ID: (Mouse) 16011
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.