Invitrogen
Manufacturer Code:MA530516
Catalog # PIMA530516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F7D12H3 |
Immunogen | Recombit Human IGFBP4 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IBP-4; IGF-binding protein 4; IGFBP4 insulin-like growth factor binding protein 4 BP-4 HT29-IGFBP IBP4 IGFBP-4 IGF-binding protein 4 insulin-like growth factor-binding protein 4; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: BP-4; HT29-IGFBP; IBP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692
Entrez Gene ID: (Human) 3487
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.