Invitrogen
Manufacturer Code:PA525925
Catalog # PIPA525925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 96-124 amino acids from the central region of human IGFBP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Band 3A; BP-4 HT29-IGFBP IBP4 IGFBP-4 IBP-4 IGF-binding protein 4 insulin-like growth factor-binding protein 4; differentially expressed in B16F10 2; IBP-4; IGF-binding protein 4; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: AI875747; BP-4; Deb2; HT29-IGFBP; IBP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692, (Mouse) P47879
Entrez Gene ID: (Human) 3487, (Mouse) 16010
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.