Invitrogen
Manufacturer Code:PA5115390
Catalog # PIPA5115390
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human IBP4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI875747 Band 3A BP-4 Deb2 differentially expressed in B16F10 2 HT29-IGFBP IBP4 IBP-4 IGF-binding protein 4 IGFBP IGFBP4 IGF-BP4 IGFBP-4 Insulin like growth factor binding protein insulin like growth factor binding protein 4 insulin-like growth factor binding protein 4 insulin-like growth factor-binding protein 4; IBP-4; IGF-binding protein 4; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: BP-4; HT29-IGFBP; IBP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692
Entrez Gene ID: (Human) 3487
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.