Proteintech
Manufacturer Code:185001AP150UL
Catalog # 50-173-2167
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | IGFBP4 Fusion Protein Ag13347 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BP 4 HT29 IGFBP IBP 4 IBP4 IGF binding protein 4 IGFBP 4 IGFBP4; IBP-4; IGF-binding protein 4; IGFBP-4; insulin-like growth factor binding protein 4; Insulin-like growth factor-binding protein 4
Gene Aliases: BP-4; HT29-IGFBP; IBP4; IGF-BP4; IGFBP-4; IGFBP4
UniProt ID: (Human) P22692, (Rat) P21744
Entrez Gene ID: (Human) 3487, (Rat) 360622
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.