Invitrogen
Manufacturer Code:MA530515
Catalog # PIMA530515
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 10 |
Immunogen | Recombit Human IGFBP3/IGFBP-3 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: acid stable subunit of the 140 K IGF complex; binding protein 29; binding protein 53; growth hormone-dependent binding protein; IBP-3; IGF-binding protein 3; IGFBP-3; insulin-like growth factor binding protein 3; Insulin-like growth factor-binding protein 3; tcag7.703 BP-53 IBP3
Gene Aliases: BP-53; IBP3; IGFBP3
UniProt ID: (Human) Q2V509
Entrez Gene ID: (Human) 3486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.