Invitrogen
Manufacturer Code:PA579452
Catalog # PIPA579452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat IGFBP3 recombit protein (Position: A29-K268). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP-3; IGF-binding protein 3; IGFBP-3; insulin-like growth factor-binding protein (IGF-BP3); Insulin-like growth factor-binding protein 3; Insulin-like growth factor-binding protein 3 IBP-3 IGF-binding protein 3 IGFBP-3 Igfbp3 Igfbp-3
Gene Aliases: AI649005; IGF-BP3; IGFBP-3; Igfbp3; IGgfbp3
UniProt ID: (Mouse) P47878, (Rat) P15473
Entrez Gene ID: (Mouse) 16009, (Rat) 24484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.