Abnova
Manufacturer Code:PAB7567
Catalog # 89-133-372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human IGFBP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of IGFBP3.
For Research Use Only
Protein Aliases: acid stable subunit of the 140 K IGF complex; binding protein 29; binding protein 53; BP-53; growth hormone-dependent binding protein; IBP-3; IBP3; IGF-binding protein 3; IGFBP-3; insulin-like growth factor binding protein 3; Insulin-like growth factor-binding protein 3
Gene Aliases: BP-53; IBP3; IGFBP3
UniProt ID: (Human) Q2V509
Entrez Gene ID: (Human) 3486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.