Proteintech
Manufacturer Code:666441IG150UL
Catalog # 50-173-7062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F10D1 |
Immunogen | IGFBP2 Fusion Protein Ag8421 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BP2 IBP 2 IBP2 IGF binding protein 2 IGF BP53 IGFBP 2 IGFBP2; IBP-2; IGF-binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; Insulin-like growth factor-binding protein 2
Gene Aliases: AI255832; BP2; IBP-2; IBP2; IGF-BP53; Igfbp-2; IGFBP2; mIGFBP-2
UniProt ID: (Human) P18065, (Mouse) P47877
Entrez Gene ID: (Human) 3485, (Mouse) 16008
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.