Invitrogen
Manufacturer Code:702196
Catalog # 70-219-6
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1H2L9 |
Immunogen | Protein corresponding to Human IGFBP2 (aa 130-325) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI255832 BP2 BRL-BP IBP2 IBP-2 IGF binding protein 2 IGF-binding protein 2 IGFBP IGFBP2 Igfbp-2 IGF-BP53 ILGFBPA Insulin like growth factor binding protein insulin like growth factor binding protein 2 insulin-like growth factor binding protein 2 insulin-like growth factor binding protein 2 36kDa insulin-like growth factor-binding protein 2 mIGFBP-2 OTTHUMP00000207753; IBP-2; IGF-binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36kDa; Insulin-like growth factor-binding protein 2
Gene Aliases: BP2; IBP2; IGF-BP53; IGFBP2
UniProt ID: (Human) P18065
Entrez Gene ID: (Human) 3485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.