Invitrogen
Manufacturer Code:MA531156
Catalog # PIMA531156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 106 |
Immunogen | Recombit Mouse IGFBP-2/IGFBP2 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: IBP-2; IBP2 IGF-BP53; IGF-binding protein 2; Insulin-like growth factor-binding protein 2
Gene Aliases: AI255832; IBP-2; Igfbp-2; Igfbp2; mIGFBP-2
UniProt ID: (Mouse) P47877
Entrez Gene ID: (Mouse) 16008
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.