Invitrogen
Manufacturer Code:PA579450
Catalog # PIPA579450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat IGFBP2 recombit protein (Position: E35-Q304). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRL-BP; IBP-2; IGF-binding protein 2; Insulin-like growth factor-binding protein 2; Insulin-like growth factor-binding protein 2 IBP-2 IGF-binding protein 2 IGFBP-2 BRL-BP Igfbp2 Igfbp-2
Gene Aliases: AI255832; IBP-2; IGFBP-2; Igfbp2; ILGFBPA; mIGFBP-2
UniProt ID: (Mouse) P47877, (Rat) P12843
Entrez Gene ID: (Mouse) 16008, (Rat) 25662
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.