OriGene
Manufacturer Code:TA808606
Catalog # 50-168-2254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 26-259 of human IGFBP1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AFBP IBP1 IGF-BP25 PP12 hIGFBP-1; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; IBP-1; IGF-binding protein 1; IGFBP-1; insulin-like growth factor binding protein 1; Insulin-like growth factor-binding protein 1; Placental protein 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBP1; PP12
UniProt ID: (Human) P08833
Entrez Gene ID: (Human) 3484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.