Invitrogen
Manufacturer Code:PA576684
Catalog # PIPA576684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human IGFBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFBPAlpha pregcy associated endometrial globulinAmniotic fluid binding proteinBinding protein 25Binding protein 26Binding protein 28Growth hormone independent binding proteinhIGFBP 1hIGFBP1IBP 1IBP-1IBP1IBP1&lowbarHUMANIGF binding protein 1IGF BP25IGF-binding protein 1IGFBP 1IGFBP-1IGFBP1Insulin like growth factor binding protein 1Insulin-like growth factor-binding protein 1Placental protein 12PP 12PP12; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; IBP-1; IGF-binding protein 1; IGFBP-1; insulin-like growth factor binding protein 1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); Insulin-like growth factor-binding protein 1; Placental protein 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBA; Igfbp-1; IGFBP1; PP12
UniProt ID: (Human) P08833, (Rat) P21743, (Mouse) P47876
Entrez Gene ID: (Human) 3484, (Rat) 25685, (Mouse) 16006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.