Abnova
Manufacturer Code:PAB27615
Catalog # 89-129-125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 156-170 of human IGFBP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of IGFBP1.
For Research Use Only
Protein Aliases: AFBP; alpha-pregnancy-associated endometrial globulin; amniotic fluid binding protein; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; hIGFBP-1; IBP-1; IBP1; IGF-binding protein 1; IGF-BP25; IGFBP-1; insulin-like growth factor binding protein 1; Insulin-like growth factor-binding protein 1; Placental protein 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBP1; PP12
UniProt ID: (Human) P08833
Entrez Gene ID: (Human) 3484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.