Novus Biologicals
Manufacturer Code:NBP122453F
Catalog # NBP122453F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0097-12C23 |
Immunogen | Mouse recombinant protein of IGFBP-5 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: IBP-5; IBP5IBP-5 IGF-binding protein 5 IGFBP-5 insulin-like growth factor binding protein 5 insulin-like growth factor-binding protein 5; IGF-binding protein 5; IGFBP-5; insulin-like growth factor binding protein 5; Insulin-like growth factor-binding protein 5
Gene Aliases: IBP5; IGFBP5
UniProt ID: (Human) P24593
Entrez Gene ID: (Human) 3488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.