R&D Systems
Manufacturer Code:AF578SP
Catalog # AF578SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-5 Val15-Glu271 Accession # Q3UQV0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP-5; IBP5IBP-5 IGF-binding protein 5 IGFBP5 IGFBP-5 insulin-like growth factor binding protein 5 insulin-like growth factor-binding protein 5; IGF-binding protein 5; IGFBP-5; insulin-like growth factor binding protein 5; Insulin-like growth factor-binding protein 5
Gene Aliases: IBP5; IGFBP5
UniProt ID: (Human) P24593
Entrez Gene ID: (Human) 3488
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.