Novus Biologicals
Manufacturer Code:NBP1224520.025MG
Catalog # NB018432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0096-11B16 |
Immunogen | Mouse recombinant protein of IGFBP-3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: acid stable subunit of the 140 K IGF complex; acid stable subunit of the 140 K IGF complex binding protein 29 binding protein 53 growth hormone-dependent binding protein IBP-3 IBP3BP-53 IGF-binding protein 3 IGFBP-3 insulin-like growth factor binding protein 3 insulin-like growth factor-binding protein 3; binding protein 29; binding protein 53; growth hormone-dependent binding protein; IBP-3; IGF-binding protein 3; IGFBP-3; insulin-like growth factor binding protein 3; Insulin-like growth factor-binding protein 3
Gene Aliases: BP-53; IBP3; IGFBP3
UniProt ID: (Human) Q2V509
Entrez Gene ID: (Human) 3486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.