R&D Systems
Manufacturer Code:MAB775
Catalog # MAB775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 138202 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-3 Pro22-Gln291 (Arg250Gln Gln259Arg and Arg271Pro) Accession # P47878 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: acid stable subunit of the 140 K IGF complex; acid stable subunit of the 140 K IGF complex binding protein 29 binding protein 53 growth hormone-dependent binding protein IBP-3 IBP3BP-53 IGF-binding protein 3 IGFBP3 IGFBP-3 insulin-like growth factor binding protein 3 insulin-like growth factor-binding protein 3; binding protein 29; binding protein 53; growth hormone-dependent binding protein; IBP-3; IGF-binding protein 3; IGFBP-3; insulin-like growth factor binding protein 3; Insulin-like growth factor-binding protein 3
Gene Aliases: BP-53; IBP3; IGFBP3
UniProt ID: (Human) Q2V509
Entrez Gene ID: (Human) 3486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.