Novus Biologicals
Manufacturer Code:NBP271011B
Catalog # NBP271011B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6C7 |
Immunogen | Recombinant protein fragment corresponding to amino acids 26-259 of human IGFBP-1 (NP_000587) produced in E.coli. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha-pregnancy-associated endometrial globulin; alpha-pregnancy-associated endometrial globulin amniotic fluid binding protein binding protein-25 binding protein-26 binding protein-28 growth hormone independent-binding protein hIGFBP-1 IBP1 IBP-1 IGF-binding protein 1 IGFBP-1 IGF-BP25 insulin-like growth factor binding protein 1 insulin-like growth factor-binding protein 1 Placental protein 12 PP12AFBP; amniotic fluid binding protein; binding protein-25; binding protein-26; binding protein-28; growth hormone independent-binding protein; IBP-1; IGF-binding protein 1; IGFBP-1; insulin-like growth factor binding protein 1; Insulin-like growth factor-binding protein 1; Placental protein 12; PP12
Gene Aliases: AFBP; hIGFBP-1; IBP1; IGF-BP25; IGFBP1; PP12
UniProt ID: (Human) P08833
Entrez Gene ID: (Human) 3484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.