Invitrogen
Manufacturer Code:PA579447
Catalog # PIPA579447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse IGFBP1 recombit protein (Position: A26-N272). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IBP-1; IGF-binding protein 1; IGFBP-1; INSULIN-LIKE GROWTH FACTOR BINDING PROTEIN 1 PRECURSOR (IGFBP-1) (IBP-1) (IGF-BINDING PROTEIN 1); Insulin-like growth factor-binding protein 1; Insulin-like growth factor-binding protein 1 IBP-1 IGF-binding protein 1 IGFBP-1 Igfbp1 Igfbp-1; placental protein 12
Gene Aliases: IBP1; IGF-BP25; IGFBA; Igfbp-1; Igfbp1
UniProt ID: (Rat) P21743, (Mouse) P47876
Entrez Gene ID: (Rat) 25685, (Mouse) 16006
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.