Invitrogen
Manufacturer Code:PA517144
Catalog # PIPA517144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the sequence of human IMP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Coding region determinant-binding protein; CRD-BP; IGF II mRNA binding protein 1 IGF-II mRNA-binding protein 1 IGF2 mRNA-binding protein 1 VICKZ family member 1 ZBP-1 coding region determit-binding protein insulin-like growth factor 2 mRNA-binding protein 1 zip code-binding protein 1 zipcode-binding protein 1; IGF-II mRNA-binding protein 1; IGF2 mRNA-binding protein 1; insulin-like growth factor 2 mRNA binding protein 1 deltaN CRDBP; Insulin-like growth factor 2 mRNA-binding protein 1; VICKZ family member 1; ZBP-1; Zipcode-binding protein 1
Gene Aliases: CRD-BP; CRDBP; IGF2BP1; IMP-1; IMP1; VICKZ1; ZBP1
UniProt ID: (Human) Q9NZI8
Entrez Gene ID: (Human) 10642
Molecular Function: RNA binding protein enzyme modulator mRNA processing factor mRNA splicing factor nucleic acid binding ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.