Invitrogen
Manufacturer Code:MA512247
Catalog # PIMA512247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M23 |
Immunogen | Purified human IGF-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IGF-I; IGF-IA IGF-IB MGFinsulin-like growth factor 1 insulin-like growth factor IA insulin-like growth factor IB mechano growth factor somatomedin-C; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; Mechano growth factor; MGF; Somatomedin; Somatomedin-C
Gene Aliases: C730016P09Rik; IBP1; Igf-1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019, (Rat) P08025, (Mouse) P05017
Entrez Gene ID: (Human) 3479, (Rat) 24482, (Mouse) 16000
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.