Invitrogen
Manufacturer Code:PA130828
Catalog # PIPA130828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli expressed human IGF-I |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IGF-I; Insulin Like Growth Factor 1 IGFI; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; Mechano growth factor; MGF; Somatomedin-C
Gene Aliases: IBP1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019
Entrez Gene ID: (Human) 3479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.