Abnova
Manufacturer Code:PAB9527
Catalog # 89-134-204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Igf1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Igf1.
For Research Use Only
Protein Aliases: C730016P09Rik; Igf-1; IGF-I; Insulin-like growth factor I; Somatomedin
Gene Aliases: C730016P09Rik; Igf-1; Igf-I; Igf1
UniProt ID: (Mouse) P05017
Entrez Gene ID: (Mouse) 16000
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.