Abnova
Manufacturer Code:PAB9526
Catalog # 89-134-203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human IGF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant IGF1.
For Research Use Only
Protein Aliases: IGF-I; IGFI; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; Mechano growth factor; MGF; Somatomedin-C
Gene Aliases: IBP1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019
Entrez Gene ID: (Human) 3479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.