Novus Biologicals
Manufacturer Code:NB500464UV
Catalog # NB500464UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MEM-238 |
Immunogen | Recombinant Vaccinia virus encoding CD222. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 300 kDa mannose 6-phosphate receptor; 300 kDa mannose 6-phosphate receptor cation-independent mannose-6 phosphate receptor cation-independent mannose-6-phosphate receptor CD222 CD222 antigen CI Man-6-P receptor CIMPR CI-MPR IGF-II receptor insulin-like growth factor 2 receptorM6P/IGF2R Insulin-like growth factor II receptor M6P/IGF2 receptor M6PR M6P-R MPR 300 MPR1 MPRIM6PR; Cation-independent mannose-6-phosphate receptor; CD222; CI Man-6-P receptor; IGF-II receptor; Insulin-like growth factor 2 receptor; Insulin-like growth factor II receptor; M6P/IGF2 receptor; M6P/IGF2R; MPR 300
Gene Aliases: CD222; CI-M6PR; CIMPR; IGF2R; M6P-R; M6P/IGF2R; MPR 300; MPR1; MPR300; MPRI
UniProt ID: (Human) P11717
Entrez Gene ID: (Human) 3482
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.