Novus Biologicals
Manufacturer Code:NBP234409H
Catalog # NBP234409H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SPM406 |
Immunogen | Purified human IGF-1 protein |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: IBP1 IGF1A IGFI IGF-I IGF-IA IGF-IB insulin-like growth factor 1 insulin-like growth factor 1 (somatomedin C) insulin-like growth factor I insulin-like growth factor IA insulin-like growth factor IB Mechano growth factor MGF somatomedin-C; IGF-I; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; Mechano growth factor; MGF; Somatomedin-C
Gene Aliases: IBP1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019
Entrez Gene ID: (Human) 3479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.