R&D Systems
Manufacturer Code:MAB2913SP
Catalog # MAB2913SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 997211 |
Immunogen | <em>E. coli</em>-derived recombinant human IGF-I/IGF-1 Gly49-Ala118 Accession # P05019 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rat Monoclonal Antibody
Protein Aliases: IBP1 IGF1 IGF-1 IGF1A IGFI IGF-I IGF-IA IGF-IB insulin-like growth factor 1 insulin-like growth factor 1 (somatomedin C) insulin-like growth factor I insulin-like growth factor IA insulin-like growth factor IB Mechano growth factor MGF Somatomedin A Somatomedin C Somatomedin-C; IGF-I; insulin-like growth factor 1 (somatomedin C); Insulin-like growth factor I; insulin-like growth factor IB; Mechano growth factor; MGF; Somatomedin-C
Gene Aliases: IBP1; IGF-I; IGF1; IGFI; MGF
UniProt ID: (Human) P05019
Entrez Gene ID: (Human) 3479
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.