Novus Biologicals
Manufacturer Code:NBP271009AF405
Catalog # N271009A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4F6 |
Immunogen | Full length recombinant protein of human IGBP1 (NP_001542) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ALPHA-4 B cell signal transduction molecule alpha 4 bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ) B-cell signal transduction molecule alpha 4 CD79a-binding protein 1 IBP1 immunoglobulin (CD79A) binding protein 1 immunoglobulin-binding protein 1 Protein alpha-4 protein phosphatase 2A regulatory subunit alpha-4 Renal carcinoma antigen NY-REN-16; B cell signal transduction molecule alpha 4; B-cell signal transduction molecule alpha 4; bA351K23.1 (immunoglobulin binding protein 1 (CD79A) ); CD79a-binding protein 1; Immunoglobulin-binding protein 1; Protein alpha-4; Protein phosphatase 2/4/6 regulatory subunit; protein phosphatase 2A, regulatory subunit alpha-4; Renal carcinoma antigen NY-REN-16
Gene Aliases: ALPHA-4; IBP1; IGBP1
UniProt ID: (Human) P78318
Entrez Gene ID: (Human) 3476
Molecular Function: defense/immunity protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.