Invitrogen
Manufacturer Code:PA570318
Catalog # PIPA570318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse IFT80 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATD2 WDR56 WD repeat domain 56 WD repeat-containing protein 56 intraflagellar transport protein 80 homolog; intraflagellar transport 80 homolog; Intraflagellar transport protein 80 homolog; intraflagellar transport protein 80i-like protein; WD repeat domain 56; WD repeat-containing protein 56
Gene Aliases: 4921524P20Rik; ATD2; IFT80; KIAA1374; mKIAA1374; RGD1312006; SRTD2; WDR56
UniProt ID: (Human) Q9P2H3, (Mouse) Q8K057, (Rat) Q66HB3
Entrez Gene ID: (Dog) 488132, (Human) 57560, (Bovine) 513583, (Pig) 100515277, (Rabbit) 100348500, (Horse) 100049959, (Mouse) 68259, (Rat) 295106
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.