Invitrogen
Manufacturer Code:PA557439
Catalog # PIPA557439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human IFT80 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATD2 WDR56 WD repeat domain 56 WD repeat-containing protein 56 intraflagellar transport protein 80 homolog; intraflagellar transport 80 homolog; Intraflagellar transport protein 80 homolog; WD repeat domain 56; WD repeat-containing protein 56
Gene Aliases: ATD2; IFT80; KIAA1374; SRTD2; WDR56
UniProt ID: (Human) Q9P2H3
Entrez Gene ID: (Human) 57560
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.