Abnova
Manufacturer Code:PAB15842
Catalog # 89-117-904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 134 amino acids of mouse Ift80. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Ift80.
For Research Use Only
Protein Aliases: 4921524P20Rik; intraflagellar transport 80 homolog; Intraflagellar transport protein 80 homolog; mKIAA1374; WD repeat domain 56; WD repeat-containing protein 56; Wdr56
Gene Aliases: 4921524P20Rik; Ift80; Kiaa1374; mKIAA1374; Wdr56
UniProt ID: (Mouse) Q8K057
Entrez Gene ID: (Mouse) 68259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.