Invitrogen
Manufacturer Code:PA5112312
Catalog # PIPA5112312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human IFT20/intraflagellar transport 20 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610009H04Rik AU015496 hIFT20 Ift20 intraflagellar transport 20 intraflagellar transport 20 homolog Intraflagellar transport protein 20 homolog intraflagellar transport protein IFT20 mIFT20 RGD1309400 uterine protein; hIFT20; intraflagellar transport 20 homolog; Intraflagellar transport protein 20 homolog; intraflagellar transport protein IFT20
Gene Aliases: IFT20
UniProt ID: (Human) Q8IY31
Entrez Gene ID: (Human) 90410
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.