Invitrogen
Manufacturer Code:PA541317
Catalog # PIPA541317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human IFT140 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: intraflagellar transport 140; intraflagellar transport 140 homolog; Intraflagellar transport protein 140 homolog; T-complex expressed gene 5; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2
Gene Aliases: AI661311; c305C8.4; c380F5.1; gs114; IFT140; KIAA0590; mKIAA0590; MZSDS; SRTD9; Tce5; WDTC2
UniProt ID: (Human) Q96RY7, (Mouse) E9PY46
Entrez Gene ID: (Rabbit) 100355837, (Human) 9742, (Horse) 100065433, (Mouse) 106633, (Rat) 100362124
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.